missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Stearate, NF, BP, JP, 4-5%, Spectrum™ Chemical

Catalog No. 18604118
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604118 45 kg Poly Bottle
18604113 500 g Poly Bottle
18604115 2.5 kg Poly Bottle
18604114 6 x 500 g Poly Bottle
18604116 4 x 2.5 kg Poly Bottle
18604117 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18604118 Supplier Spectrum Chemical Mfg Cor Supplier No. MA13045KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MA130, 557-04-0

Spectrum™ Chemical Magnesium Stearate, NF, BP, JP is used as an anti-adherent in the manufacture of of medical capsules, tablets, and powders. Spectrum Chemical manufactured NF, BP, JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 557-04-0
Molecular Formula C36H70MgO4
Molecular Weight (g/mol) 591.26
InChI Key HQKMJHAJHXVSDF-UHFFFAOYSA-L
IUPAC Name magnesium(2+) dioctadecanoate
SMILES [Mg++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O

Specifications

Physical Form Powder, Needles or Crystals
CAS Max % 100%
Quantity 45 kg
Loss on Drying 6%
Packaging Poly Bottle
Percent Purity 4 to 5%
Grade BP/EP/JP/NF
Chemical Name or Material Magnesium Stearate