missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Stearate, FCC, Spectrum™ Chemical

Catalog No. p-7158578
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
4 x 2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604108 500 g Poly Pail
18604110 2.5 kg Poly Pail
18604109 6 x 500 g Poly Pail
18604111 4 x 2.5 kg Poly Pail
18604112 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18604108 Supplier Spectrum Chemical Mfg Cor Supplier No. M1184500GM
Only null left

M1184, 557-04-0

Spectrum™ Chemical Magnesium Stearate, FCC is often used as an anti-adherent since it has lubricating properties, preventing ingredients from sticking to manufacturing equipment during the compression of chemical powders into solid tablets; magnesium stearate is the most commonly used lubricant for tablets. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 557-04-0
Molecular Formula C36H70MgO4
Molecular Weight (g/mol) 591.26
InChI Key HQKMJHAJHXVSDF-UHFFFAOYSA-L
IUPAC Name magnesium(2+) dioctadecanoate
SMILES [Mg++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O

Specifications

CAS Max % 1
Quantity 500 g
Grade FCC
Packaging Poly Pail