missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Stearate, FCC, Spectrum™ Chemical

Catalog No. 18604112
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
4 x 2.5 kg
45 kg
Packaging:
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604112 45 kg Poly Pail
18604108 500 g Poly Pail
18604110 2.5 kg Poly Pail
18604109 6 x 500 g Poly Pail
18604111 4 x 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18604112 Supplier Spectrum Chemical Mfg Cor Supplier No. M118445KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

M1184, 557-04-0

Spectrum™ Chemical Magnesium Stearate, FCC is often used as an anti-adherent since it has lubricating properties, preventing ingredients from sticking to manufacturing equipment during the compression of chemical powders into solid tablets; magnesium stearate is the most commonly used lubricant for tablets. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 557-04-0
Molecular Formula C36H70MgO4
Molecular Weight (g/mol) 591.26
InChI Key HQKMJHAJHXVSDF-UHFFFAOYSA-L
IUPAC Name magnesium(2+) dioctadecanoate
SMILES [Mg++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O

Specifications

CAS Max % 1
Physical Form Powder, Needles or Crystals
Quantity 45 kg
Percent Purity 6.8 to 8.3%
Grade FCC
Packaging Poly Pail
Chemical Name or Material Magnesium Stearate