missing translation for 'onlineSavingsMsg'
Learn More

Magnesium stearate, 3.8-5.0% Mg

Catalog No. AC240760250
Change view
Click to view available options
Quantity:
25 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC240760250 25 g Plastic bottle
AC240760010 1 kg Plastic bottle
AC240760050 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC240760250 Supplier Thermo Scientific Chemicals Supplier No. 240760250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 557-04-0 | C36H70MgO4 | 591.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 557-04-0
Molecular Formula C36H70MgO4
Molecular Weight (g/mol) 591.27
InChI Key HQKMJHAJHXVSDF-UHFFFAOYSA-L
Synonym magnesium stearate, magnesium octadecanoate, magnesium distearate, synpro 90, octadecanoic acid, magnesium salt, dibasic magnesium stearate, petrac mg 20nf, ns-m salt, sm-p, stearic acid, magnesium salt
PubChem CID 11177
ChEBI CHEBI:9254
IUPAC Name magnesium;octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Mg+2]

Specifications

Melting Point 130.0°C to 140.0°C
Color White
Loss on Drying 6% (1g, 105°C) max.
Infrared Spectrum Authentic
Packaging Plastic bottle
Linear Formula Mg(C18H35O2)2
Quantity 25 g
Merck Index 15,5754
Solubility Information Solubility in water: insoluble. Other solubilities: decomposes in diluted acids,practically insoluble in alcohols and ether
Formula Weight 591.27
Percent Purity 3.8-5% Mg
Physical Form Fine Powder
Chemical Name or Material Magnesium stearate
Show More Show Less
EINECSNumber 209-150-3

RUO – Research Use Only