missing translation for 'onlineSavingsMsg'
Learn More

Magnesium stearate, 3.8-5.0% Mg

CAS: 557-04-0 | C36H70MgO4 | 591.27 g/mol

Supplier:  Thermo Scientific Chemicals 240760050

Quantity 5 kg
Packaging Plastic drum

 View more versions of this product

Catalog No. AC240760050


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

557-04-0
591.27
magnesium stearate, magnesium octadecanoate, magnesium distearate, synpro 90, octadecanoic acid, magnesium salt, dibasic magnesium stearate, petrac mg 20nf, ns-m salt, sm-p, stearic acid, magnesium salt
CHEBI:9254
CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Mg+2]
C36H70MgO4
HQKMJHAJHXVSDF-UHFFFAOYSA-L
11177
magnesium;octadecanoate

Specifications

Magnesium stearate
130.0°C to 140.0°C
6% (1g, 105°C) max.
Plastic drum
Mg(C18H35O2)2
15,5754
Solubility in water: insoluble. Other solubilities: decomposes in diluted acids,practically insoluble in alcohols and ether
CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Mg+2]
591.27
CHEBI:9254
3.8-5% Mg
557-04-0
White
Authentic
C36H70MgO4
5 kg
magnesium stearate, magnesium octadecanoate, magnesium distearate, synpro 90, octadecanoic acid, magnesium salt, dibasic magnesium stearate, petrac mg 20nf, ns-m salt, sm-p, stearic acid, magnesium salt
HQKMJHAJHXVSDF-UHFFFAOYSA-L
magnesium;octadecanoate
11177
591.27
Fine Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 209-150-3

SDS
Documents

Documents

Promotions

Promotions