missing translation for 'onlineSavingsMsg'
Learn More

Magnesium slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog No. AA4329809
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4329809 10 g
AA4329818 50 g
AA4329830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4329809 Supplier Thermo Scientific Chemicals Supplier No. 04329809

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-95-4
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]

Specifications

Quantity 10 g
Odor Odorless
Weight ≈0.35g/slug
Form Slug
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Magnesium slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length
Hazard Category H228-H252-H261
Precautionary Statement P210-P223-P231+P232-P235+P410-P240-P241-P280-P335+P334-P370+P378s-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: III
EINECSNumber 231-104-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only