missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Powder, Particle size about 0.06-0.3mm, MilliporeSigma™
Specifications
Specifications
| Appearance | Light gray solid |
| Quantity | 1 kg |
| Odor | Odorless |
| Packaging | Metal can |
| Form | Solid |
| UN Number | UN 1418 |
| Formula Weight | 24.31. (molar mass) |
| Chemical Name or Material | Magnesium |
Safety and Handling
| Hazard Category | H250: Catches fire spontaneously if exposed to air. H260: In contact with water releases flammable gases which may ignite spontaneously. |