missing translation for 'onlineSavingsMsg'
Learn More

Magnesium phthalocyanine, tech. 90%

Catalog No. p-7050556
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3860103 1 g
AA3860106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3860103 Supplier Thermo Scientific Chemicals Supplier No. 03860103
Only null left

CAS: 1661-03-6 | C32H18MgN8O | 554.856 g/mol

Magnesium phthalocyanine is used in preparing membrane sensors for cyanide ions. Also used in preparation of membrane sensors for cyanide ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1661-03-6
Molecular Formula C32H18MgN8O
Molecular Weight (g/mol) 554.856
MDL Number MFCD00041953
InChI Key HTNHYCHFBSVBID-UHFFFAOYSA-N
Synonym magnesium phthalocyanine
PubChem CID 91872988
SMILES C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.O.[Mg+2]

Specifications

Quantity 1 g
Solubility Information Soluble in concentrated sulfuric acid.
Formula Weight 536.84
Percent Purity 90%
Grade Technical
Physical Form Powder
Chemical Name or Material Magnesium phthalocyanine
Hazard Category H250-H314-H335
Precautionary Statement P210-P222-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
EINECSNumber 216-766-6
RTECSNumber OM4205000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only