missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Phosphate, Dibasic, Trihydrate, FCC, 96%, Spectrum™ Chemical
MA127, 7782-75-4, MgHPO4·3H2O
$567.70 - $4371.37
Chemical Identifiers
| CAS | 7782-75-4 |
|---|---|
| Molecular Formula | H7MgO7P |
| Molecular Weight (g/mol) | 174.33 |
| MDL Number | MFCD00150390 |
| InChI Key | OKIWLDVQGKRUNR-UHFFFAOYSA-L |
| IUPAC Name | magnesium(2+) trihydrate hydrogen phosphate |
| SMILES | O.O.O.[Mg++].OP([O-])([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18604095
|
Spectrum Chemical Mfg Cor
MA127500GM |
500 g | Poly Bottle |
Each for $567.70
|
|
||||
|
18604096
|
Spectrum Chemical Mfg Cor
MA12725KG |
2.5 kg | Poly Pail |
Each for $1,352.17
|
|
||||
|
18604097
|
Spectrum Chemical Mfg Cor
MA12712KG |
12 kg | Poly Pail |
Each for $4,371.37
|
|
||||
Description
Spectrum™ Chemical Magnesium Phosphate, Dibasic, Trihydrate, FCC is listed on the FDA′s Generally recognized as safe, or GRAS, list of substances. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 7782-75-4 | |
| 174.33 | |
| OKIWLDVQGKRUNR-UHFFFAOYSA-L | |
| O.O.O.[Mg++].OP([O-])([O-])=O |
| H7MgO7P | |
| MFCD00150390 | |
| magnesium(2+) trihydrate hydrogen phosphate |
Specifications
| 7782-75-4 | |
| 500 g | |
| MFCD00150390 | |
| O.O.O.[Mg++].OP([O-])([O-])=O | |
| 174.33 | |
| FCC | |
| Poly Bottle |
| 100% | |
| H7MgO7P | |
| OKIWLDVQGKRUNR-UHFFFAOYSA-L | |
| magnesium(2+) trihydrate hydrogen phosphate | |
| 96% | |
| 29.0-36% |