missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Perchlorate Hydrate (about 83% Mg(ClO4)2], For Elemental Analysis, MilliporeSigma™

Catalog No. m1058750500
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1058750500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1058750500 Supplier Technidata Supplier No. 1.05875.0500

Chemical Identifiers

CAS 64010-42-0
Molecular Formula Cl2H2MgO9
Molecular Weight (g/mol) 241.212
InChI Key KKFFNQXCJMNXHI-UHFFFAOYSA-L
Synonym magnesium perchlorate hydrate, magnesium diperchlorate hydrate, 2clo4.mg.h2o, magnesium 2+ ion hydrate diperchlorate, magnesium 2+ hydrate diperchlorate, magnesium perchlorate hydrate, puriss. p.a calc. based on dry substance, kt
PubChem CID 6915996
IUPAC Name magnesium;diperchlorate;hydrate
SMILES O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2]

Specifications

Physical Form Solid
Quantity 500 g
UN Number UN 1475
Odor Odorless
Packaging Metal can
Appearance White solid
Chemical Name or Material Magnesium perchlorate hydrate
Hazard Category H272: May intensify fire; oxidiser.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.