missing translation for 'onlineSavingsMsg'
Learn More

Magnesium perchlorate hexahydrate, 99% (metals basis)

CAS: 13446-19-0 | Cl2H12MgO14 | 331.287 g/mol

Supplier:  Thermo Scientific Chemicals 01163530

 View more versions of this product

Catalog No. AA1163530


Only null left
Explore available promotions

Description

Description

Commercially available magnesium perchlorate is reported as an extremely efficient catalyst for the synthesis of α-aminophosphonates. Magnesium perchlorate has been found to be an efficient catalyst for the synthesis of imines and phenylhydrazones by the reaction of carbonyl compounds with amines and phenylhydrazine in high yields at room temperatures and in short times.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Magnesium perchlorate hexahydrate
13446-19-0
Crystals
99% (Metals basis)
Cl2H12MgO14
UN1475
magnesium perchlorate hexahydrate, acmc-20akl9, 2clo4.mg.6h2o, ksc491a3f, magnesium 2+ ion hexahydrate diperchlorate, magnesium 2+ hexahydrate diperchlorate
MGBDNCBRZIMSDZ-UHFFFAOYSA-L
magnesium;diperchlorate;hexahydrate
16211969
99%
Hygroscopic
185°C to 190°C
White
250 g
(metals basis)
MFCD00149778
14,5678
Insoluble in water.
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2]
331.287
331.33 (223.23 Anhydrous)
Odorless

Chemical Identifiers

13446-19-0
331.287
MGBDNCBRZIMSDZ-UHFFFAOYSA-L
16211969
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2]
Cl2H12MgO14
MFCD00149778
magnesium perchlorate hexahydrate, acmc-20akl9, 2clo4.mg.6h2o, ksc491a3f, magnesium 2+ ion hexahydrate diperchlorate, magnesium 2+ hexahydrate diperchlorate
magnesium;diperchlorate;hexahydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: MAGNESIUM PERCHLORATE

EINECSNumber : 233-108-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only