missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Perchlorate, Desiccant, ACS, Spectrum™ Chemical

Catalog No. 18611444
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
Packaging:
Amber Glass Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611444 125 g Amber Glass Bottle
18611445 500 g Amber Glass Bottle
18611446 2.5 kg Poly Pail
18611447 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18611444 Supplier Spectrum Chemical Mfg Cor Supplier No. M1060125GM

Chemical Identifiers

CAS 10034-81-8
Molecular Formula Cl2MgO8
Molecular Weight (g/mol) 223.20
InChI Key MPCRDALPQLDDFX-UHFFFAOYSA-L
IUPAC Name magnesium(2+) diperchlorate
SMILES [Mg++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

CAS Max % 1
Quantity 125 g
Grade ACS
Loss on Drying 0.08
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.