missing translation for 'onlineSavingsMsg'
Learn More

Magnesium perchlorate, ACS

Catalog No. AA1163636
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1163636 500 g
AA1163622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1163636 Supplier Thermo Scientific Chemicals Supplier No. 01163636

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Magnesium Perchlorate, Anhydrous, Cl2MgO8, CAS Number-10034-81-8, magnesium perchlorate, anhydrous, anhydrous magnesium perchlorate, anhydrone, magnesium perchlorate, magnesium diperchlorate, hsdb 661, dehydrite, perchlorate de magnesium french, perchloric acid, magnesium salt, unii-7n77z541yf, 500g | CAS: 10034-81-8 | Cl2MgO8 | 223.197 g/mol

Magnesium perchlorate is commonly utilized as a potent oxidizing agent. It is used as dehydrating agent in organic synthesis. It is practiced in many domains like pharma industry, pyrotechnics industry,in rechargeable batteries and it is a versatile tool in organic synthesis. It possesses photoelectronic and optical properties. Thus it can be used in photo electronic and optoelectronic devices. It is applied as a strong drying agent for gases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10034-81-8
Molecular Formula Cl2MgO8
Molecular Weight (g/mol) 223.197
MDL Number MFCD00011102
InChI Key MPCRDALPQLDDFX-UHFFFAOYSA-L
Synonym anhydrone, magnesium perchlorate, dehydrite, perchloric acid, magnesium salt, anhydrous magnesium perchlorate, unii-7n77z541yf, hsdb 661, magnesium diperchlorate, magnesium perchlorate, anhydrous, perchlorate de magnesium french
PubChem CID 24840
IUPAC Name magnesium;diperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2]

Specifications

Melting Point 251°C (decomposition)
Physical Form Granules
Quantity 500 g
Beilstein EM>5,
Solubility Information Magnesium perchlorate is soluble in water,ethanol.
Formula Weight 223.21
Grade ACS Reagent
Sensitivity Hygroscopic
Density 2.21 g/mL
Chemical Name or Material Magnesium Perchlorate, Anhydrous, For analysis ACS
Hazard Category H272-H315-H319-H335
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: II
EINECSNumber 233-108-3
RTECSNumber SC8925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only