missing translation for 'onlineSavingsMsg'
Learn More

Magnesium perchlorate, ACS reagent, desiccant

Catalog No. AC413465000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC413465000 500 g Plastic bottle
AC413461000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC413465000 Supplier Thermo Scientific Chemicals Supplier No. 413465000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Magnesium perchlorate, Cl2MgO8, CAS Number-10034-81-8, anhydrous magnesium perchlorate, unii-7n77z541yf, perchloric acid, magnesium salt, perchlorate de magnesium french, dehydrite, magnesium perchlorate, anhydrous, magnesium perchlorate, anhydrone, hsdb 661, magnesium diperchlorate, 500g, White | CAS: 10034-81-8 | Cl2MgO8 | 223.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent, Oxidation reagent

Chemical Identifiers

CAS 10034-81-8
Molecular Formula Cl2MgO8
Molecular Weight (g/mol) 223.21
MDL Number MFCD00011102
InChI Key MPCRDALPQLDDFX-UHFFFAOYSA-L
Synonym anhydrone, magnesium perchlorate, dehydrite, perchloric acid, magnesium salt, anhydrous magnesium perchlorate, unii-7n77z541yf, hsdb 661, magnesium diperchlorate, magnesium perchlorate, anhydrous, perchlorate de magnesium french
PubChem CID 24840
IUPAC Name magnesium;diperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2]

Specifications

Melting Point 250.0°C
Color White
Physical Form Crystalline Powder, Flakes, and Chunks
Quantity 500 g
Loss on Drying 8% max. (190°C)
Packaging Plastic bottle
Linear Formula Mg(ClO4)2
Merck Index 15, 5742
Solubility Information Solubility in water: 993g/L (25°C). Other solubilities: very soluble in alcohol
Formula Weight 223.21
Grade ACS Reagent
Chemical Name or Material Magnesium perchlorate, Desiccant
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Take any precaution to avoid mixing with combustibles.
IF ON SKIN: Wash
EINECSNumber 233-108-3

RUO – Research Use Only