missing translation for 'onlineSavingsMsg'
Learn More

Magnesium nitride, 99.6% (metals basis)

Catalog No. aa4194618
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4194618 50 g
AA4194609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4194618 Supplier Thermo Scientific Chemicals Supplier No. 04194618
Only null left

CAS: 12057-71-5

The use of magnesium nitride as a convenient source of ammonia allows a direct transformation of esters to primary amides. Methyl, ethyl, isopropyl, and tert-butyl esters are converted to the corresponding carboxamides in good yields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12057-71-5
MDL Number MFCD00016205

Specifications

Melting Point 800°C (decomposition)
Physical Form Powder
Quantity 50 g
UN Number UN3178
Solubility Information Reacts with water.
Formula Weight 100.95
Percent Purity 99.6%
Sensitivity Moisture sensitive
Density 2.71 g/mL
Assay (metals basis)
Chemical Name or Material Magnesium nitride
Show More Show Less
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
EINECSNumber 235-022-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only