missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Nitrate Hexahydrate, Honeywell Fluka™

Catalog No. p-7071920
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002174 500 g
6002172 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002174 Supplier Honeywell Chemicals Supplier No. 237175500G
Only null left

ACS Reagent, 99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 13446-18-9
Molecular Formula H12MgN2O12
Molecular Weight (g/mol) 256.40
MDL Number MFCD00149779
InChI Key MFUVDXOKPBAHMC-UHFFFAOYSA-N
Synonym magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty
PubChem CID 202877
IUPAC Name magnesium;dinitrate;hexahydrate

Specifications

Quantity 500 g
Linear Formula Mg(NO3)2 · 6H2O
UN Number NONH for all modes of transport
SMILES O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Formula Weight 256.41g/mol
Percent Purity 99%
Grade ACS Reagent
Density Vapor Desnity: 6 (vs air)
Chemical Name or Material Magnesium Nitrate Hexahydrate
EINECSNumber 233-826-7
RTECSNumber OM3756000