missing translation for 'onlineSavingsMsg'
Learn More

Magnesium nitrate hexahydrate, extra pure

Catalog No. p-4813530
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic Bottle
Plastic Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC447540010 1 kg Plastic Bottle
AC447540050 5 kg Plastic Drum
AC447540250 25 kg Fibre drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC447540010 Supplier Thermo Scientific Chemicals Supplier No. 447540010
Only null left

CAS: 13446-18-9 | H12MgN2O12 | 256.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13446-18-9
Molecular Formula H12MgN2O12
Molecular Weight (g/mol) 256.40
MDL Number MFCD00149779
InChI Key MFUVDXOKPBAHMC-UHFFFAOYSA-N
Synonym magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty
PubChem CID 202877
IUPAC Name magnesium;dinitrate;hexahydrate
SMILES O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Boiling Point 330.0°C
Melting Point 89.0°C
Quantity 1 kg
Linear Formula Mg(NO3)2·6H2O
Flash Point 93.3°C
Merck Index 15,5738
Solubility Information Solubility in water: 420g/L (20°C). Other solubilities: freely soluble in alcohol, liquid nh3
Formula Weight 256.4
Grade Extra Pure
Packaging Plastic Bottle
Chemical Name or Material Magnesium Nitrate Hexahydrate
Show More Show Less

RUO – Research Use Only