missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Nitrate, Hexahydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical

Catalog No. 18611434
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611434 125 g Amber Glass Bottle
18611435 500 g Amber Glass Bottle
18611436 2.5 kg Poly Pail
18611437 12 kg Poly Pail
18611438 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18611434 Supplier Spectrum Chemical Mfg Cor Supplier No. M1050125GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 13446-18-9
Molecular Formula H12MgN2O12
Molecular Weight (g/mol) 256.40
InChI Key MFUVDXOKPBAHMC-UHFFFAOYSA-N
IUPAC Name magnesium(2+) hexahydrate dinitrate
SMILES O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

CAS Max % 1
pH 5.0 to 8.2
Quantity 125 g
Percent Purity 98 to 102%
Grade ACS
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.