missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Magnesium nitrate hexahydrate, for biochemistry

CAS: 13446-18-9 | H12MgN2O12 | 256.40 g/mol

$677.03 - $677.03

Chemical Identifiers

CAS 13446-18-9
Molecular Formula H12MgN2O12
Molecular Weight (g/mol) 256.40
MDL Number MFCD00149779
InChI Key MFUVDXOKPBAHMC-UHFFFAOYSA-N
Synonym magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty
PubChem CID 202877
IUPAC Name magnesium;dinitrate;hexahydrate
SMILES O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC446830025
SDS View Documents
Thermo Scientific Chemicals
446830025
2.5 kg Plastic bottle
Each for $677.03
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13446-18-9
256.40
MFUVDXOKPBAHMC-UHFFFAOYSA-N
202877
O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
H12MgN2O12
MFCD00149779
magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty
magnesium;dinitrate;hexahydrate

Specifications

330.0°C
13446-18-9
H12MgN2O12
MFCD00149779
15,5738
Solubility in water: 420g/L (20°C). Other solubilities: freely soluble in alcohol, liquid nh3
O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
256.40
256.4
Plastic bottle
89.0°C
2.5 kg
Mg(NO3)2·6H2O
93.3°C
magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty
MFUVDXOKPBAHMC-UHFFFAOYSA-N
magnesium;dinitrate;hexahydrate
202877
Biochemical
Magnesium nitrate hexahydrate
Videos
Safety and Handling

Safety and Handling

Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only