missing translation for 'onlineSavingsMsg'
Learn More

Magnesium niobium oxide, 99.9% (metals basis)

Catalog No. AA3937330
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3937330 250 g
AA3937309 10 g
AA3937318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3937330 Supplier Thermo Scientific Chemicals Supplier No. 03937330
Only null left

CAS: 12163-26-7 | Mg2Nb2O7 | 346.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12163-26-7
Molecular Formula Mg2Nb2O7
Molecular Weight (g/mol) 346.42
MDL Number MFCD00210631
InChI Key CWMQWEHVVUJOBW-UHFFFAOYSA-N
Synonym Magnesium niobate
IUPAC Name diniobium(5+) dimagnesium(2+) heptaoxidandiide
SMILES [O--].[O--].[O--].[O--].[O--].[O--].[O--].[Mg++].[Mg++].[Nb+5].[Nb+5]

Specifications

Physical Form Powder
Quantity 250 g
Solubility Information Insoluble in water.
Formula Weight 306.11
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Magnesium niobium oxide
EINECSNumber 235-310-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only