missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium meso-tetraphenylporphine monohydrate
CAS: 14640-21-2 | C44H28MgN4 | 637.04 g/mol
$270.10 - $900.30
Chemical Identifiers
| CAS | 14640-21-2 |
|---|---|
| Molecular Formula | C44H28MgN4 |
| Molecular Weight (g/mol) | 637.04 |
| MDL Number | MFCD00049815 |
| InChI Key | XEHJAWQTIIXDON-QHNUXLDVSA-N |
| Synonym | magnesium meso-tetraphenylporphine hydrate |
| PubChem CID | 131675734 |
| IUPAC Name | magnesium;2,7,12,17-tetraphenylporphyrin-21,22-diide |
| SMILES | C1=CC=C(C=C1)C2=CC3=CC4=C(C=C([N-]4)C=C5C(=CC(=N5)C=C6C(=CC(=N6)C=C2[N-]3)C7=CC=CC=C7)C8=CC=CC=C8)C9=CC=CC=C9.[Mg+2] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 14640-21-2 | |
| 637.04 | |
| XEHJAWQTIIXDON-QHNUXLDVSA-N | |
| 131675734 | |
| C1=CC=C(C=C1)C2=CC3=CC4=C(C=C([N-]4)C=C5C(=CC(=N5)C=C6C(=CC(=N6)C=C2[N-]3)C7=CC=CC=C7)C8=CC=CC=C8)C9=CC=CC=C9.[Mg+2] |
| C44H28MgN4 | |
| MFCD00049815 | |
| magnesium meso-tetraphenylporphine hydrate | |
| magnesium;2,7,12,17-tetraphenylporphyrin-21,22-diide |
Specifications
| 14640-21-2 | |
| MFCD00049815 | |
| magnesium meso-tetraphenylporphine hydrate | |
| C1=CC=C(C=C1)C2=CC3=CC4=C(C=C([N-]4)C=C5C(=CC(=N5)C=C6C(=CC(=N6)C=C2[N-]3)C7=CC=CC=C7)C8=CC=CC=C8)C9=CC=CC=C9.[Mg+2] | |
| 637.04 | |
| 655.05 (637.04 Anhydrous) | |
| Magnesium meso-tetraphenylporphine monohydrate |
| C44H28MgN4 | |
| 0.5 g | |
| XEHJAWQTIIXDON-QHNUXLDVSA-N | |
| magnesium;2,7,12,17-tetraphenylporphyrin-21,22-diide | |
| 131675734 | |
| Crystalline |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only