missing translation for 'onlineSavingsMsg'
Learn More

Magnesium meso-tetraphenylporphine monohydrate

Catalog No. AA3931602
Change view
Click to view available options
Quantity:
0.5 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3931602 0.5 g
AA3931604 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3931602 Supplier Thermo Scientific Chemicals Supplier No. 03931602

CAS: 14640-21-2 | C44H28MgN4 | 637.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14640-21-2
Molecular Formula C44H28MgN4
Molecular Weight (g/mol) 637.04
MDL Number MFCD00049815
InChI Key XEHJAWQTIIXDON-QHNUXLDVSA-N
Synonym magnesium meso-tetraphenylporphine hydrate
PubChem CID 131675734
IUPAC Name magnesium;2,7,12,17-tetraphenylporphyrin-21,22-diide
SMILES C1=CC=C(C=C1)C2=CC3=CC4=C(C=C([N-]4)C=C5C(=CC(=N5)C=C6C(=CC(=N6)C=C2[N-]3)C7=CC=CC=C7)C8=CC=CC=C8)C9=CC=CC=C9.[Mg+2]

Specifications

Quantity 0.5 g
Formula Weight 655.05 (637.04 Anhydrous)
Physical Form Crystalline
Chemical Name or Material Magnesium meso-tetraphenylporphine monohydrate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only