Learn More
Magnesium lactate trihydrate, 99%
CAS: 18917-96-9 | C6H16MgO9 | 256.49 g/mol
$102.38 - $291.27
Chemical Identifiers
| CAS | 18917-96-9 |
|---|---|
| Molecular Formula | C6H16MgO9 |
| Molecular Weight (g/mol) | 256.49 |
| MDL Number | MFCD00156534 |
| Synonym | magnesium lactate trihydrate, c6h10mgo6.3h2o, 2-hydroxypropanoyl oxy magnesio 2-hydroxypropanoate trihydrate |
| PubChem CID | 91668363 |
| IUPAC Name | magnesium;2-hydroxypropanoate;trihydrate |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 18917-96-9 | |
| 256.49 | |
| magnesium lactate trihydrate, c6h10mgo6.3h2o, 2-hydroxypropanoyl oxy magnesio 2-hydroxypropanoate trihydrate | |
| magnesium;2-hydroxypropanoate;trihydrate |
| C6H16MgO9 | |
| MFCD00156534 | |
| 91668363 |
Specifications
| 18917-96-9 | |
| White | |
| MFCD00156534 | |
| 14,5672 | |
| Soluble in water, Slightly soluble in alcohol | |
| CC(C(=O)[O-])O.CC(C(=O)[O-])O.O.O.O.[Mg+2] | |
| 256.49 | |
| 256.49 (202.45 Anhydrous) | |
| Powder or Lumps |
| 41°C (decomposition) | |
| C6H16MgO9 | |
| 25 g | |
| magnesium lactate trihydrate, c6h10mgo6.3h2o, 2-hydroxypropanoyl oxy magnesio 2-hydroxypropanoate trihydrate | |
| SNLQXUYQWUDJLB-UHFFFAOYSA-L | |
| magnesium;2-hydroxypropanoate;trihydrate | |
| 91668363 | |
| 99% | |
| Magnesium lactate trihydrate |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 242-671-4
RTECSNumber : OD5660000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only