missing translation for 'onlineSavingsMsg'
Learn More

Magnesium(II) Stearate, TCI America™

Catalog No. S023825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S023825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S023825G Supplier TCI America Supplier No. S023825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 557-04-0
Molecular Formula C36H70MgO4
Molecular Weight (g/mol) 591.257
MDL Number MFCD00036391
InChI Key HQKMJHAJHXVSDF-UHFFFAOYSA-L
Synonym magnesium stearate, magnesium octadecanoate, magnesium distearate, synpro 90, octadecanoic acid, magnesium salt, dibasic magnesium stearate, petrac mg 20nf, ns-m salt, sm-p, stearic acid, magnesium salt
PubChem CID 11177
ChEBI CHEBI:9254
IUPAC Name magnesium;octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Mg+2]

Specifications

Color White
Quantity 25 g
Formula Weight 591.26
Physical Form Crystalline Powder
Chemical Name or Material Magnesium(II) Stearate
EINECSNumber (2)-0611
RTECSNumber WI4390000
TSCA Yes