missing translation for 'onlineSavingsMsg'
Learn More

Magnesium(II) Gluconate Hydrate 98.0+%, TCI America™

Catalog No. G027625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G027625G 25 g
G0276500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G027625G Supplier TCI America Supplier No. G027625G

Chemical Identifiers

CAS 3632-91-5
Molecular Formula C12H22MgO14
Molecular Weight (g/mol) 414.60
MDL Number MFCD00150971
InChI Key CTUVIUYTHWPELF-IYEMJOQQSA-L
Synonym Gluconic Acid Magnesium(II) Salt
PubChem CID 91886357
IUPAC Name magnesium(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
SMILES [Mg++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

Specifications

Color White-Yellow
Physical Form Crystalline Powder
Quantity 25 g
Formula Weight 414.60
Percent Purity ≥98.0% (T)
Chemical Name or Material Magnesium(II) Gluconate Hydrate
EINECSNumber (2)-1410
RTECSNumber OM3480000
TSCA Yes