missing translation for 'onlineSavingsMsg'
Learn More

Magnesium(II) Bis(trifluoromethanesulfonyl)imide 97.0+%, TCI America™

Catalog No. M28611G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28611G 1 g
M28615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28611G Supplier TCI America Supplier No. M28611G

Chemical Identifiers

CAS 133395-16-1
Molecular Formula C4F12MgN2O8S4
Molecular Weight (g/mol) 584.576
MDL Number MFCD14156020
InChI Key DMFBPGIDUUNBRU-UHFFFAOYSA-N
Synonym bis bis trifluoromethylsulfonyl amino magnesium, magnesium bis trifluoromethanesulfonimide, magnesium ii bis trifluoromethanesulfonyl imide, magnesium ii triflimide, magnesium bistrifluoromethanesulfonimide, magnesium bis trifluoromethylsulfonyl imide, magnesium 2+ bis bistriflylimide anion, bis trifluoromethanesulfonyl imide magnesium ii salt
PubChem CID 11851070
IUPAC Name magnesium;bis(trifluoromethylsulfonyl)azanide
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Mg+2]

Specifications

Color White
Quantity 1 g
UN Number 1759
Formula Weight 584.58
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Magnesium(II) Bis(trifluoromethanesulfonyl)imide