missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium(II) 2-Ethylbutyrate 95.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 254.61 |
| Percent Purity | ≥95.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Magnesium(II) 2-Ethylbutyrate |
Safety and Handling
| TSCA | No |