missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Hydroxide, Purified, Reagents

Catalog No. 01337188
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337188 2.5 kg
01337187 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337188 Supplier Labchem Inc Supplier No. C22387002.5KG

Magnesium Hydroxide, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1309-42-8
Molecular Formula H2MgO2
Molecular Weight (g/mol) 58.32
MDL Number MFCD00011104
InChI Key VTHJTEIRLNZDEV-UHFFFAOYSA-L
Synonym Milk of Magnesia, Magnesium Dihydroxide
IUPAC Name magnesium(2+) dihydroxide
SMILES [OH-].[OH-].[Mg++]

Specifications

Boiling Point 673°C
Color Colorless/White
Physical Form Solid
Quantity 2.5 kg
Grade Pure
Packaging Poly Pail
Specific Gravity 2.36
Chemical Name or Material Magnesium Hydroxide, Purified