missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Hydroxide, Powder, USP, 95-100.5%, Spectrum™ Chemical

Catalog No. p-7157790
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
100 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604074 500 g Poly Bottle
18604077 100 lb. Fiber Drum
18604075 2.5 kg Poly Pail
18604076 12 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18604074 Supplier Spectrum Chemical Mfg Cor Supplier No. MA115500GM
Only null left

MA115, 1309-42-8, Mg(OH)2

Spectrum™ Chemical Magnesium Hydroxide, Powder, USP is used to treat symptoms caused by too much stomach acid such as heartburn, upset stomach, or indigestion. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1309-42-8
Molecular Formula H2MgO2
Molecular Weight (g/mol) 58.32
MDL Number MFCD00011104
InChI Key VTHJTEIRLNZDEV-UHFFFAOYSA-L
IUPAC Name magnesium(2+) dihydroxide
SMILES [OH-].[OH-].[Mg++]

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 95 to 100.5%
Grade USP
Loss on Drying 2%
Loss on Ignition 30.0-33%
Packaging Poly Bottle