missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Hydroxide, Powder, USP, 95-100.5%, Spectrum™ Chemical
Description
Spectrum™ Chemical Magnesium Hydroxide, Powder, USP is used to treat symptoms caused by too much stomach acid such as heartburn, upset stomach, or indigestion. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 2.5 kg |
| Percent Purity | 95 to 100.5% |
| Grade | USP |
| Loss on Drying | 2% |
| Loss on Ignition | 30.0-33% |
| Packaging | Poly Pail |