missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Hydroxide, Powder, 95.0-100.5%, Spectrum™ Chemical

Catalog No. p-7165819
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611428 500 g Poly Bottle
18611427 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18611428 Supplier Spectrum Chemical Mfg Cor Supplier No. M1048500GM
Only null left

Chemical Identifiers

CAS 1309-42-8

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 95 to 100.5%
Grade Reagent
Loss on Drying 0.02
Loss on Ignition 30.0-33.0%
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.