missing translation for 'onlineSavingsMsg'
Learn More

Magnesium hydrogen phosphate trihydrate, 98%, extra pure, Thermo Scientific Chemicals

$152.04 - $152.04

Chemical Identifiers

CAS 7782-75-4
Molecular Formula H7MgO7P
Molecular Weight (g/mol) 174.33
MDL Number MFCD00150390
InChI Key OKIWLDVQGKRUNR-UHFFFAOYSA-L
Synonym magnesium hydrogen phosphate trihydrate, dimagnesium phosphate, unii-hf539g9l3q, magnesium phosphate dibasic trihydrate, magnesium 2+ trihydrate hydrogen phosphate, magnesium 2+ ion trihydrate hydrogen phosphate, kc defense, newberyite, magnesium dibasic phosphate trihydrate, acmc-20aklg
PubChem CID 16204530
IUPAC Name magnesium;hydrogen phosphate;trihydrate
SMILES O.O.O.[Mg++].OP([O-])([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC264780010
SDS View Documents
Thermo Scientific Chemicals
264780010
1 kg Plastic bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7782-75-4
174.33
OKIWLDVQGKRUNR-UHFFFAOYSA-L
16204530
O.O.O.[Mg++].OP([O-])([O-])=O
H7MgO7P
MFCD00150390
magnesium hydrogen phosphate trihydrate, dimagnesium phosphate, unii-hf539g9l3q, magnesium phosphate dibasic trihydrate, magnesium 2+ trihydrate hydrogen phosphate, magnesium 2+ ion trihydrate hydrogen phosphate, kc defense, newberyite, magnesium dibasic phosphate trihydrate, acmc-20aklg
magnesium;hydrogen phosphate;trihydrate

Specifications

White
Powder
97.5% min. (Complexometry)
MgHPO4·3H2O
14, 5681
Solubility in water: 0.25g/L (20°C). Other solubilities: soluble in diluted acids
O.O.O.[Mg++].OP([O-])([O-])=O
174.33
174.34
Extra Pure
2.13
Magnesium hydrogen phosphate trihydrate, c.p., 98%
7782-75-4
1 kg
H7MgO7P
MFCD00150390
magnesium hydrogen phosphate trihydrate, dimagnesium phosphate, unii-hf539g9l3q, magnesium phosphate dibasic trihydrate, magnesium 2+ trihydrate hydrogen phosphate, magnesium 2+ ion trihydrate hydrogen phosphate, kc defense, newberyite, magnesium dibasic phosphate trihydrate, acmc-20aklg
OKIWLDVQGKRUNR-UHFFFAOYSA-L
magnesium;hydrogen phosphate;trihydrate
16204530
98%
Plastic bottle
2.1300g/mL
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only