Learn More
Magnesium Gluconate, USP, 98-102%, Spectrum™ Chemical
MA112, 3632-91-5, C12H22MgO14
Supplier:  Spectrum Chemical Mfg Cor MA11212KG
 View more versions of this product
Catalog No. 18604068
Only null left
Explore available promotions
Description
Description
Spectrum™ Chemical Magnesium Gluconate, USP is an essential mineral and used in treating low magnesium levels or maintaining the proper amount of magnesium in the body. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
3632-91-5
6.0-7.8
C12H22MgO14
CTUVIUYTHWPELF-IYEMJOQQSA-L
magnesium(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
98 to 102%
Poly Pail
100%
12 kg
MFCD00150971
[Mg++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
414.60
USP
Chemical Identifiers
3632-91-5
414.60
CTUVIUYTHWPELF-IYEMJOQQSA-L
[Mg++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
C12H22MgO14
MFCD00150971
magnesium(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
Product Suggestions
Product Suggestions