missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Gluconate, USP, 98-102%, Spectrum™ Chemical

Catalog No. 18604065
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604065 100 g Poly Bottle
18604066 500 g Poly Bottle
18604067 2.5 kg Poly Pail
18604068 12 kg Poly Pail
18604069 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18604065 Supplier Spectrum Chemical Mfg Cor Supplier No. MA112100GM

MA112, 3632-91-5, C12H22MgO14

Spectrum™ Chemical Magnesium Gluconate, USP is an essential mineral and used in treating low magnesium levels or maintaining the proper amount of magnesium in the body. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 3632-91-5
Molecular Formula C12H22MgO14
Molecular Weight (g/mol) 414.60
MDL Number MFCD00150971
InChI Key CTUVIUYTHWPELF-IYEMJOQQSA-L
IUPAC Name magnesium(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
SMILES [Mg++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

Specifications

pH 6.0-7.8
CAS Max % 100%
Quantity 100 g
Percent Purity 98 to 102%
Grade USP
Packaging Poly Bottle