missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Gluconate, Dihydrate, FCC, 98-102%, Spectrum™ Chemical

Catalog No. 18604062
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604062 100 g Poly Bottle
18604063 500 g Poly Bottle
18604064 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18604062 Supplier Spectrum Chemical Mfg Cor Supplier No. M1134100GM

M1134, 59625-89-7, C12H22MgO14·2H2O

Spectrum™ Chemical Magnesium Gluconate, Dihydrate, FCC is a mineral supplement used to prevent and treat low amounts of magnesium in the blood. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 59625-89-7

Specifications

CAS Max % 100%
Quantity 100 g
Percent Purity 98 to 102%
Grade FCC
Packaging Poly Bottle