missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Foil, 0.15-0.30mm Thickness, 3mm Wide, MilliporeSigma™

Catalog No. M1058120001
Change view
Click to view available options
Quantity:
1 Roll
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1058120001 1 Roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1058120001 Supplier Technidata Supplier No. 1.05812.0001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7439-95-4
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]

Specifications

Appearance Metallic solid
Quantity 1 Roll
Odor Odorless
Packaging Fibre case
Form Solid
UN Number UN 1869
Formula Weight 24.31. (molar mass)
Chemical Name or Material Magnesium