missing translation for 'onlineSavingsMsg'
Learn More

Magnesium fluoride, Puratronic™, 99.99% (metals basis)

CAS: 7783-40-6 | F2Mg | 62.30 g/mol

Supplier:  Thermo Scientific Chemicals 04075509

 View more versions of this product

Catalog No. AA4075509


Only null left
Explore available promotions

Description

Description

Used in optics, space telescopes. Most commonly used on the surfaces of optical elements as inexpensive anti-reflective coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Magnesium fluoride
7783-40-6
Powder
(metals basis)
MgF2
14,5665
ORUIBWPALBXDOA-UHFFFAOYSA-L
magnesium(2+) difluoride
62.3
Puratronic
1.35-1.38
1261°C
White
10 g
F2Mg
MFCD00011108
Soluble in water,nitric acid. Insoluble in ethanol.
[F-].[F-].[Mg++]
62.30
99.99%
Odorless
3.148 g/mL

Chemical Identifiers

7783-40-6
62.30
ORUIBWPALBXDOA-UHFFFAOYSA-L
[F-].[F-].[Mg++]
F2Mg
MFCD00011108
magnesium(2+) difluoride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 231-995-1

RTECSNumber : OM3325000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only