missing translation for 'onlineSavingsMsg'
Learn More

Magnesium fluoride hydrate, min 87% MgF2

CAS: 7783-40-6 | F2Mg | 62.30 g/mol

Supplier:  Thermo Scientific Chemicals A128300E

 View more versions of this product

Catalog No. AAA128300E


Only null left
Explore available promotions

Description

Description

Magnesium fluoride is used in ceramics, optical lenses, glass manufacturing, and single crystals for polarizing prisms. It is also used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium fluoride is used in ceramics, optical lenses, glass manufacturing, and single crystals for polarizing prisms. It is also used in organic synthesis.

Solubility
Soluble in water (87 mg/L at 18°C ).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with Strong oxidizing agents and strong acids. Store at room temperature.
Specifications

Specifications

Magnesium fluoride hydrate
2500 g
F2Mg
14,5665
ORUIBWPALBXDOA-UHFFFAOYSA-L
magnesium(2+) difluoride
62.30 (Anhydrous)
7783-40-6
≥87% MgF2
MFCD00011108
Soluble in water (87mg/L at 18°C ).
[F-].[F-].[Mg++]
62.30
Odorless

Chemical Identifiers

7783-40-6
62.30
ORUIBWPALBXDOA-UHFFFAOYSA-L
[F-].[F-].[Mg++]
F2Mg
MFCD00011108
magnesium(2+) difluoride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 231-995-1

RTECSNumber : OM3325000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only