missing translation for 'onlineSavingsMsg'
Learn More

Magnesium fluoride hydrate, min 87% MgF2

Catalog No. AAA1283036
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1283036 500 g
AAA128300E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1283036 Supplier Thermo Scientific Chemicals Supplier No. A1283036

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7783-40-6 | F2Mg | 62.30 g/mol

Magnesium fluoride is used in ceramics, optical lenses, glass manufacturing, and single crystals for polarizing prisms. It is also used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium fluoride is used in ceramics, optical lenses, glass manufacturing, and single crystals for polarizing prisms. It is also used in organic synthesis.

Solubility
Soluble in water (87 mg/L at 18°C ).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with Strong oxidizing agents and strong acids. Store at room temperature.

Chemical Identifiers

CAS 7783-40-6
Molecular Formula F2Mg
Molecular Weight (g/mol) 62.30
MDL Number MFCD00011108
InChI Key ORUIBWPALBXDOA-UHFFFAOYSA-L
IUPAC Name magnesium(2+) difluoride
SMILES [F-].[F-].[Mg++]

Specifications

Quantity 500 g
Odor Odorless
Merck Index 14,5665
Solubility Information Soluble in water (87mg/L at 18°C ).
Formula Weight 62.30 (Anhydrous)
Assay ≥87% MgF2
Chemical Name or Material Magnesium fluoride hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 231-995-1
RTECSNumber OM3325000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only