missing translation for 'onlineSavingsMsg'
Learn More

Magnesium fluoride, 99% (metals basis excluding Ca & Na), Ca+Na <1%

Catalog No. AA35807A1
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35807A1 1 kg
AA3580718 50 g
AA3580730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA35807A1 Supplier Thermo Scientific Chemicals Supplier No. 035807A1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7783-40-6 | F2Mg | 62.30 g/mol

Used in optics, space telescopes. Most commonly used on the surfaces of optical elements as inexpensive anti-reflective coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7783-40-6
Molecular Formula F2Mg
Molecular Weight (g/mol) 62.30
MDL Number MFCD00011108
InChI Key ORUIBWPALBXDOA-UHFFFAOYSA-L
IUPAC Name magnesium(2+) difluoride
SMILES [F-].[F-].[Mg++]

Specifications

Melting Point 1261°C
Color White
Physical Form Powder
Quantity 1 kg
Odor Odorless
Refractive Index 1.35-1.38
Assay Percent Range 99%, Ca+Na <1% (Metals basis excluding Ca and Na)
Density 3.148
Solubility Information Sparingly soluble in water. Slightly soluble in dilute HNO3Insoluble in ethanol.
Formula Weight 62.3
Chemical Name or Material Magnesium fluoride
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 231-995-1
RTECSNumber OM3325000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only