missing translation for 'onlineSavingsMsg'
Learn More

Magnesium fluoride, 99.9% (metals basis excluding Ca & Na), Ca+Na <1%

Catalog No. AA3639114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3639114 25 g
AA3639122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3639114 Supplier Thermo Scientific Chemicals Supplier No. 03639114

Vacuum deposition | CAS: 7783-40-6 | F2Mg | 62.30 g/mol

Magnesium fluoride is used as Vacuum deposition, anti-reflective coatings, in optics and also used in space telescopes, glass industry and ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7783-40-6
Molecular Formula F2Mg
Molecular Weight (g/mol) 62.30
MDL Number MFCD00011108
InChI Key ORUIBWPALBXDOA-UHFFFAOYSA-L
IUPAC Name magnesium(2+) difluoride
SMILES [F-].[F-].[Mg++]

Specifications

Melting Point 1261°C
Color White
Physical Form Vacuum Melted Lump
Quantity 25 g
Linear Formula MgF2
Merck Index 14,5665
Solubility Information Soluble in water(0.076g/L),nitric acid. Insoluble in ethanol.
Formula Weight 62.3
Percent Purity 99.9%
Odor Odorless
Refractive Index 1.35-1.38
Density 3.148 g/mL
Assay (metals basis excluding Ca and Na), Ca+Na <1%
Chemical Name or Material Magnesium fluoride
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 231-995-1
RTECSNumber OM3325000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only