missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Magnesium Chloride, Hexahydrate, Molecular Biology Grade, Calbiochem™,

$113.00 - $113.00

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
Synonym MgCl2
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
442611500GM
View Documents
MilliporeSigma™
442611500GM
500 g
Each for $113.00
Only null left
 
Description

Description

Hygroscopic
Specifications

Chemical Identifiers

7791-18-6
203.30
DHRRIBDTHFBPNG-UHFFFAOYSA-L
magnesium(2+) hexahydrate dichloride
Cl2H12MgO6
MFCD00149781
MgCl2
O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

7791-18-6
Cl2H12MgO6
MgCl2
DHRRIBDTHFBPNG-UHFFFAOYSA-L
magnesium(2+) hexahydrate dichloride
203.3
500 g
MFCD00149781
Easily soluble in cold water
O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
203.30
Magnesium Chloride, Hexahydrate
Videos
Safety and Handling

Safety and Handling

RTECSNumber : OM2975000

Recommended Storage : Keep container tightly closed. Keep container in a cool, well-ventilated area.

SDS
Documents

Documents