missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium chloride hexahydrate, Honeywell Fluka™
ACS Reagent, 99.0-102.0%
Supplier: Honeywell Chemicals M9272500G
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Magnesium chloride hexahydrate | |
| 500 g | |
| MgCl2 · 6H2O | |
| NONH for all modes of transport | |
| O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] | |
| 203.30 | |
| 99 to 102% |
| 7791-18-6 | |
| Cl2H12MgO6 | |
| MFCD00149781 | |
| DHRRIBDTHFBPNG-UHFFFAOYSA-L | |
| magnesium(2+) hexahydrate dichloride | |
| 203.3g/mol |
Chemical Identifiers
| 7791-18-6 | |
| 203.30 | |
| DHRRIBDTHFBPNG-UHFFFAOYSA-L | |
| O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] |
| Cl2H12MgO6 | |
| MFCD00149781 | |
| magnesium(2+) hexahydrate dichloride |
Safety and Handling
RTECSNumber : OM2975000