missing translation for 'onlineSavingsMsg'
Learn More

Magnesium chloride hexahydrate, Honeywell Fluka™

Catalog No. 6003096
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
6 x 500 g
4 x 2.5 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6003096 100 g
6003101 500 g
6003098 1 kg
6003099 2.5 kg
6003102 6 x 500 g
6003100 4 x 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 6003096 Supplier Honeywell Chemicals Supplier No. M9272100G
Only null left

ACS Reagent, 99.0-102.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Quantity 100 g
Linear Formula MgCl2 · 6H2O
UN Number NONH for all modes of transport
Formula Weight 203.3g/mol
Percent Purity 99 to 102%
Chemical Name or Material Magnesium chloride hexahydrate
RTECSNumber OM2975000