missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride Hexahydrate, Honeywell Fluka™

Catalog No. p-7071800
Change view
Click to view available options
Quantity:
2.5 kg
6 x 2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001975 2.5 kg
60044779 6 x 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001975 Supplier Honeywell Chemicals Supplier No. 131522.5KG
Only null left

Puriss., meets analytical specification of BP, FCC, European Pharmacopoeia, E511, ≤0.0001% Al, 99-101%,

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Quantity 2.5 kg
Linear Formula MgCl2 · 6H2O
UN Number NONH for all modes of transport
Formula Weight 203.3g/mol
Percent Purity 99 to 101%
Grade puriss.
Chemical Name or Material Magnesium Chloride Hexahydrate, Meets analytical specification of Ph. Eur.BP, FCC
EINECSNumber 232-094-6
RTECSNumber OM2975000