missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Chloride, Hexahydrate, Flake, FCC, 99-105%, Spectrum™ Chemical
MA111, 7791-18-6, MgCl2·6H2O
Supplier: Spectrum Chemical Mfg Cor MA11125KGCS4
Description
Spectrum™ Chemical Magnesium Chloride, Hexahydrate, Flake, FCC is used as a nutritional supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 7791-18-6 | |
| 4 x 2.5 kg | |
| MFCD00149781 | |
| O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] | |
| 203.30 | |
| Poly Pail |
| 100% | |
| Cl2H12MgO6 | |
| DHRRIBDTHFBPNG-UHFFFAOYSA-L | |
| magnesium(2+) hexahydrate dichloride | |
| 99 to 105% |
Chemical Identifiers
| 7791-18-6 | |
| 203.30 | |
| DHRRIBDTHFBPNG-UHFFFAOYSA-L | |
| O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] |
| Cl2H12MgO6 | |
| MFCD00149781 | |
| magnesium(2+) hexahydrate dichloride |