missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride Hexahydrate, (Crystalline),MP Biomedicals

Catalog No. ICN19469880
Change view
Click to view available options
:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ICN19469880 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19469880 Supplier MP Biomedicals Inc Supplier No. 0219469880
Only null left

Specifications

CAS 7791-18-6
Physical Form Crystalline
Quantity 100 g
Molecular Formula Cl2H12MgO6
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
IUPAC Name magnesium(2+) hexahydrate dichloride
Molecular Weight (g/mol) 203.30
Formula Weight 203.3
Chemical Name or Material Magnesium Chloride Hexahydrate
Show More Show Less
Recommended Storage Room Temperature