missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride, Hexahydrate, ACS, 99.0 to 101.0%, MilliporeSigma™

Catalog No. M1058335000
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1058335000 5 kg
M1058330250 250 g
M1058331000 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1058335000 Supplier Technidata Supplier No. 1.05833.5000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
Synonym Magnesium dichloride
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Color Gray-White to White
Physical Form Solid
Quantity 5 kg
Assay Percent Range 99.0 to 101.0%
Linear Formula MgCl2 · 6H2O
Formula Weight 23.3
Grade ACS/EP/ISO/Reagent
Density 1.57g/cm3 (20°C)
Chemical Name or Material Magnesium Chloride, Hexahydrate, EMSURE™, For Analysis
Recommended Storage Store without restrictions