missing translation for 'onlineSavingsMsg'
Learn More

Magnesium chloride hexahydrate, 99%, for analysis

Catalog No. p-3734880
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic Bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC197530010 1 kg Plastic Bottle
AC197530050 5 kg Plastic drum
AC197530250 25 kg Fibre drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC197530010 Supplier Thermo Scientific Chemicals Supplier No. 197530010
Only null left

CAS: 7791-18-6 | Cl2H12MgO6 | 203.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Melting Point 117.0°C
Color White
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 98.5 to 101.5% (Complexometry)
Linear Formula MgCl2·6H2O
Fieser 10,251; 15,196
Merck Index 15, 5726
Solubility Information Solubility in water: 2350g/L (20°C). Other solubilities: soluble in alcohol
Formula Weight 203.31
Percent Purity 99%
Grade Analytical
Packaging Plastic Bottle
Chemical Name or Material Magnesium chloride hexahydrate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only