missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Chloride Hexahydrate, 99.0-101.0%, (Crystalline/ACS Reagent Grade), MP Biomedicals
Specifications
Specifications
| Physical Form | Crystalline |
| Quantity | 1 kg |
| Assay Percent Range | 99.0-101.0% |
| Molecular Formula | MgCl2 · 6H2O |
| Formula Weight | 203.3 |
| Grade | ACS Reagent |
| Chemical Name or Material | Magnesium Chloride Hexahydrate |
Safety and Handling
| Recommended Storage | Room Temperature |